accessibility menu, dialog, popup

UserName

CAS RN 160434-49-1

CAS RN 160434-49-1

IUPAC Name: (2E)-3-(3-bromo-4-fluorophenyl)prop-2-enoate
Synonyms: (2E)-3-(3-bromo-4-fluorophenyl)prop-2-enoate
Molecular Weight (g/mol): 244.04
Molecular Formula: C9H5BrFO2
InChi Key: ZNIGVADAKXOMQH-DUXPYHPUSA-M
SMILES: [O-]C(=O)\C=C\C1=CC=C(F)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results