accessibility menu, dialog, popup

UserName

CAS RN 1603-79-8

IUPAC Name: ethyl 2-oxo-2-phenylacetate
Synonyms: ethyl 2-oxo-2-phenylacetate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: QKLCQKPAECHXCQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)