accessibility menu, dialog, popup

UserName

CAS RN 1603-40-3

CH3NH2N

CAS RN 1603-40-3

IUPAC Name: 3-methylpyridin-2-amine
Synonyms: 2-pyridinamine, 3-methyl-
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: RGDQRXPEZUNWHX-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1N
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)