accessibility menu, dialog, popup

UserName

CAS RN 16024-82-1

CH3OONCS

CAS RN 16024-82-1

IUPAC Name: methyl 2-isothiocyanatobenzoate
Synonyms: methyl 2-isothiocyanatobenzoate
Molecular Weight (g/mol): 193.22
Molecular Formula: C9H7NO2S
InChi Key: UNXVHBOJSCWVCD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1N=C=S
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)