accessibility menu, dialog, popup

UserName

H3CNNNNCH3CH2CH3H2CH3CHOOHOOFeCl(Z)(Z)(Z)(Z)

CAS RN 16009-13-5

IUPAC Name: iron(3+) 4,20-bis(2-carboxyethyl)-9,14-diethenyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide
Synonyms: hemin
Molecular Weight (g/mol): 616.5
Molecular Formula: C34H32FeN4O4
InChi Key: GGIDWJQWCUJYRY-UHFFFAOYSA-L
SMILES: [Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (6)
Quantity 
  • (4)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (5)
  • (6)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)