accessibility menu, dialog, popup

UserName

CAS RN 1600-27-7

HgCH3OOCH3OO

CAS RN 1600-27-7

IUPAC Name: mercury(2+) diacetate
Synonyms: mercury(II) acetate
Molecular Weight (g/mol): 318.68
Molecular Formula: C4H6HgO4
InChi Key: BRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES: [Hg++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (1)
  • (3)
Grade 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (9)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)