CAS RN 159600-61-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-[4-[2-(2-Pyridinyloxy)propoxy]phenoxy]phenol, TRC
CAS: 159600-61-0 Molecular Formula: C20H19NO4 Molecular Weight (g/mol): 337.37 Synonym: Pyriproxyfen-4'-hydroxy,Phenol, 4-[4-[2-(2-pyridinyloxy)propoxy]phenoxy]- (9CI, ACI),4-[4-[2-(2-Pyridinyloxy)propoxy]phenoxy]phenol IUPAC Name: 4-[4-(2-pyridin-2-yloxypropoxy)phenoxy]phenol SMILES: CC(COc1ccc(Oc2ccc(O)cc2)cc1)Oc3ccccn3
2
4-OH-Pyriproxyfen (4'-OH-Pyr) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality