accessibility menu, dialog, popup

UserName

CAS RN 1594-58-7

H2NNOHN(Z)

CAS RN 1594-58-7

IUPAC Name: (Z)-N'-hydroxypyridine-3-carboximidamide
Synonyms: (Z)-N'-hydroxypyridine-3-carboximidamide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: AQBMQGDKWIPBRF-UHFFFAOYSA-N
SMILES: N\C(=N/O)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)