accessibility menu, dialog, popup

UserName

CAS RN 159326-68-8

IUPAC Name: pyrrolo[2,1-f][1,2,4]triazin-4-amine
Synonyms: pyrrolo[2,1-f][1,2,4]triazin-4-amine
Molecular Weight (g/mol): 134.14
Molecular Formula: C6H6N4
InChi Key: VSPXQZSDPSOPRO-UHFFFAOYSA-N
SMILES: NC1=NC=NN2C=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results