accessibility menu, dialog, popup

UserName

NPN

CAS RN 15909-92-9

IUPAC Name: 3-[(2-cyanoethyl)(phenyl)phosphanyl]propanenitrile
Synonyms: 3-[(2-cyanoethyl)(phenyl)phosphanyl]propanenitrile
Molecular Weight (g/mol): 216.22
Molecular Formula: C12H13N2P
InChi Key: LIYGDNBFEBKBBZ-UHFFFAOYSA-N
SMILES: N#CCCP(CCC#N)C1=CC=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results