accessibility menu, dialog, popup

UserName

OOSNH2

CAS RN 15884-65-8

IUPAC Name: 2H-1,3-benzodioxole-5-carbothioamide
Synonyms: 2H-1,3-benzodioxole-5-carbothioamide
Molecular Weight (g/mol): 181.21
Molecular Formula: C8H7NO2S
InChi Key: YHXXBQMLJHUUJU-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results