accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OPrONOOONOOONOO

CAS RN 15878-77-0

IUPAC Name: praseodymium(3+) hexahydrate trinitrate
Synonyms: praseodymium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 435.01
Molecular Formula: H12N3O15Pr
InChi Key: LXXCECZPOWZKLC-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)