accessibility menu, dialog, popup

UserName

CAS RN 15872-42-1

H3COOHO

CAS RN 15872-42-1

IUPAC Name: 4-(heptyloxy)benzoic acid
Synonyms: 4-(heptyloxy)benzoic acid
Molecular Weight (g/mol): 236.31
Molecular Formula: C14H20O3
InChi Key: ZRVIYEJYXIDATJ-UHFFFAOYSA-N
SMILES: CCCCCCCOC1=CC=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results