accessibility menu, dialog, popup

UserName

CAS RN 15855-06-8

CH3ONOHOCl

CAS RN 15855-06-8

IUPAC Name: 2-chloro-6-methoxypyridine-4-carboxylic acid
Synonyms: 2-chloro-6-methoxypyridine-4-carboxylic acid
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: PJQBTHQTVJMCFX-UHFFFAOYSA-N
SMILES: COC1=NC(Cl)=CC(=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)