accessibility menu, dialog, popup

UserName

CAS RN 15854-87-2

IUPAC Name: 4-iodopyridine
Synonyms: 4-iodopyridine
Molecular Weight (g/mol): 205
Molecular Formula: C5H4IN
InChi Key: RTLUPHDWSUGAOS-UHFFFAOYSA-N
SMILES: IC1=CC=NC=C1
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)