accessibility menu, dialog, popup

UserName

HOBOHFFFFF

CAS RN 1582-24-7

IUPAC Name: (2,3,4,5,6-pentafluorophenyl)boronic acid
Synonyms: 2,3,4,5,6-pentafluorophenylboronic acid
Molecular Weight (g/mol): 211.88
Molecular Formula: C6H2BF5O2
InChi Key: VASOMTXTRMYSKD-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results