accessibility menu, dialog, popup

UserName

CAS RN 158063-54-8

IUPAC Name: 4-(trifluoromethyl)pyridine-3-carbothioamide
Synonyms: 4-(trifluoromethyl)pyridine-3-carbothioamide
Molecular Weight (g/mol): 206.19
Molecular Formula: C7H5F3N2S
InChi Key: HOPAEPDXFOSQMZ-UHFFFAOYSA-N
SMILES: NC(=S)C1=C(C=CN=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results