accessibility menu, dialog, popup

UserName

CAS RN 15804-32-7

CH3OOCH3OOCH3OOAu

CAS RN 15804-32-7

IUPAC Name: gold(3+) triacetate
Synonyms: gold(3+) triacetate
Molecular Weight (g/mol): 374.1
Molecular Formula: C6H9AuO6
InChi Key: OTCKNHQTLOBDDD-UHFFFAOYSA-K
SMILES: [Au+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results