accessibility menu, dialog, popup

UserName

CAS RN 1578-63-8

IUPAC Name: (2S)-2-fluoro-2-phenylacetate
Synonyms: (2S)-2-fluoro-2-phenylacetate
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H6FO2
InChi Key: ATPPNMLQNZHDOG-ZETCQYMHSA-M
SMILES: [O-]C(=O)[C@@H](F)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results