accessibility menu, dialog, popup

UserName

CAS RN 157730-74-0

OS

CAS RN 157730-74-0

IUPAC Name: 4-(thiophen-3-yl)benzaldehyde
Synonyms: 4-(thiophen-3-yl)benzaldehyde
Molecular Weight (g/mol): 188.24
Molecular Formula: C11H8OS
InChi Key: MGPZYZLJJSEFLU-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results