CAS RN 157590-59-5
CAS RN 157590-59-5
IUPAC Name:
4-chloro-5-(trifluoromethyl)benzene-1,2-diamine
Synonyms:
4-chloro-5-(trifluoromethyl)benzene-1,2-diamine
Molecular Weight (g/mol):
210.58
Molecular Formula:
C7H6ClF3N2
InChi Key:
BUSGYQISMPLVIJ-UHFFFAOYSA-N
SMILES:
NC1=C(N)C=C(C(Cl)=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Chloro-5-trifluoromethyl-o-phenylenediamine, 95%, Thermo Scientific™
CAS: 157590-59-5 Molecular Formula: C7H6ClF3N2 Molecular Weight (g/mol): 210.584 MDL Number: MFCD00119550 InChI Key: BUSGYQISMPLVIJ-UHFFFAOYSA-N Synonym: 4-chloro-5-trifluoromethyl benzene-1,2-diamine,2-amino-4-chloro-5-trifluoromethyl aniline,4-chloro-5-trifluoromethyl-o-phenylenediamine,2-amino-4-chloro-5-trifluoromethyl phenylamine,2-chloro-4,5-diaminobenzotrifluoride,5-chloro-4-trifluoromethyl benzene-1,2-diamine,1,2-benzenediamine, 4-chloro-5-trifluoromethyl,aminochlorotrifluoromethylphenylamine,3,4-diamino-6-chlorobenzotrifluoride PubChem CID: 2773497 IUPAC Name: 4-chloro-5-(trifluoromethyl)benzene-1,2-diamine SMILES: C1=C(C(=CC(=C1N)N)Cl)C(F)(F)F