accessibility menu, dialog, popup

UserName

ONHNNH2HBr

CAS RN 157415-17-3

IUPAC Name: morpholine-4-carboximidamide hydrobromide
Synonyms: morpholine-4-carboximidamide hydrobromide
Molecular Weight (g/mol): 210.08
Molecular Formula: C5H12BrN3O
InChi Key: IOHSDHMCZXCESZ-UHFFFAOYSA-N
SMILES: Br.NC(=N)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results