accessibility menu, dialog, popup

UserName

HOOOHN

CAS RN 15733-89-8

IUPAC Name: 2-oxo-1,2-dihydroquinoline-4-carboxylic acid
Synonyms: 2-oxocinchoninic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: MFSHNFBQNVGXJX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results