accessibility menu, dialog, popup

UserName

CAS RN 15681-05-7

MgClClClK

CAS RN 15681-05-7

IUPAC Name: magnesium(2+) potassium trichloride
Synonyms: magnesium(2+) potassium trichloride
Molecular Weight (g/mol): 169.75
Molecular Formula: Cl3KMg
InChi Key: JNZGLUUWTFPBKG-UHFFFAOYSA-K
SMILES: [Mg++].[Cl-].[Cl-].[Cl-].[K+]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results