accessibility menu, dialog, popup

UserName

CAS RN 15629-92-2

PPNiClClPhPhPhPh

CAS RN 15629-92-2

IUPAC Name: nickel(2+) [3-(diphenylphosphaniumyl)propyl]diphenylphosphanium dichloride
Synonyms: nickel(2+) [3-(diphenylphosphaniumyl)propyl]diphenylphosphanium dichloride
Molecular Weight (g/mol): 544.06
Molecular Formula: C27H28Cl2NiP2
InChi Key: ZBQUMMFUJLOTQC-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].[Ni++].C(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)C[PH+](C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)