CAS RN 156243-64-0
IUPAC Name:
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
Synonyms:
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol):
260.99
Molecular Formula:
C7H2BrF5
InChi Key:
QPJKIRNIIXIPIE-UHFFFAOYSA-N
SMILES:
FC1=CC(Br)=CC(F)=C1C(F)(F)F
1
–
2
of
2
results
2
4-Bromo-2,6-difluorobenzotrifluoride, 98%
CAS: 156243-64-0 Molecular Formula: C7H2BrF5 Molecular Weight (g/mol): 260.989 MDL Number: MFCD01631616 InChI Key: QPJKIRNIIXIPIE-UHFFFAOYSA-N PubChem CID: 23450919 IUPAC Name: 5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene SMILES: C1=C(C=C(C(=C1F)C(F)(F)F)F)Br