accessibility menu, dialog, popup

UserName

CAS RN 15604-36-1

PPClClPt

CAS RN 15604-36-1

IUPAC Name: platinum(2+) bis(triphenylphosphane) dichloride
Synonyms: platinum(2+) bis(triphenylphosphine) dichloride
Molecular Weight (g/mol): 790.57
Molecular Formula: C36H30Cl2P2Pt
InChi Key: XAFJSPPHVXDRIE-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pt++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results