accessibility menu, dialog, popup

UserName

CAS RN 155640-85-0

OOCuOOCF3CF3CF3CF3

CAS RN 155640-85-0

IUPAC Name: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) copper hydrate
Synonyms: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) copper hydrate
Molecular Weight (g/mol): 497.68
Molecular Formula: C10H6CuF12O5
InChi Key: DBDCQCFMFYPMNY-UHFFFAOYSA-N
SMILES: O.[Cu].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (5)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results