accessibility menu, dialog, popup

UserName

NaCH3BH3CH3CH3C

CAS RN 15523-24-7

IUPAC Name: sodium tetraethylboranuide
Synonyms: sodium tetraethylboranuide
Molecular Weight (g/mol): 150.05
Molecular Formula: C8H20BNa
InChi Key: SZSBMTRYJRHYNI-UHFFFAOYSA-N
SMILES: [Na+].CC[B-](CC)(CC)CC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)