accessibility menu, dialog, popup

UserName

CAS RN 15489-90-4

FeNNNNOHOOH2CCH2CH3H3COHHOH3CCH3

CAS RN 15489-90-4

IUPAC Name: λ⁵-iron(5+) 5,9-bis(2-carboxylatoethyl)-14,19-diethenyl-4,10,15,20-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide hydrate
Synonyms: λ⁵-iron(5+) 5,9-bis(2-carboxylatoethyl)-14,19-diethenyl-4,10,15,20-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide hydrate
Molecular Weight (g/mol): 632.5
Molecular Formula: C34H32FeN4O5
InChi Key: XLKTVQRBSKAXKW-UHFFFAOYSA-J
SMILES: O.[Fe+5].CC1=C(CCC([O-])=O)C2=CC3=NC(=CC4=C(C)C(C=C)=C([N-]4)C=C4N=C(C=C1[N-]2)C(C=C)=C4C)C(C)=C3CCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results