CAS RN 154594-15-7
IUPAC Name:
N-[4-(4H-1,2,4-triazol-4-yl)phenyl]acetamide
Synonyms:
N-[4-(1,2,4-triazol-4-yl)phenyl]acetamide
Molecular Weight (g/mol):
202.22
Molecular Formula:
C10H10N4O
InChi Key:
RBAFMDFJFDEJBG-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=C(C=C1)N1C=NN=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4'-(1,2,4-Triazol-4-yl)acetanilide, 99%, Thermo Scientific™
CAS: 154594-15-7 Molecular Formula: C10H10N4O Molecular Weight (g/mol): 202.217 MDL Number: MFCD01075653 InChI Key: RBAFMDFJFDEJBG-UHFFFAOYSA-N Synonym: n-4-1,2,4-triazol-4-yl phenyl acetamide,4'-1,2,4-triazol-4-yl acetanilide,n-4-4h-1,2,4-triazol-4-yl phenyl acetamide,acetamide, n-4-4h-1,2,4-triazol-4-yl phenyl,4-1,2,4-triazol-4-yl acetanilide,4-4-acetamidophenyl-1,2,4-triazole,4-4-acetylamino phenyl-4h-1,2,4-triazole,acetamide,n-4-4h-1,2,4-triazol-4-yl phenyl PubChem CID: 4086562 IUPAC Name: N-[4-(1,2,4-triazol-4-yl)phenyl]acetamide SMILES: CC(=O)NC1=CC=C(C=C1)N2C=NN=C2