CAS RN 154590-34-8
CAS RN 154590-34-8
IUPAC Name:
tert-butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate
Synonyms:
tert-butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol):
325.34
Molecular Formula:
C15H20FN3O4
InChi Key:
YOLVWCABLVHASL-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1CCN(CC1)C1=C(F)C=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™
CAS: 154590-34-8 Molecular Formula: C15H20FN3O4 Molecular Weight (g/mol): 325.34 MDL Number: MFCD08443959 InChI Key: YOLVWCABLVHASL-UHFFFAOYSA-N PubChem CID: 16203508 IUPAC Name: tert-butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F