accessibility menu, dialog, popup

UserName

CAS RN 154259-25-3

H3COBrFFF

CAS RN 154259-25-3

IUPAC Name: 1-[3-bromo-5-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[3-bromo-5-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 267.05
Molecular Formula: C9H6BrF3O
InChi Key: BVCLQCAMSFVBFX-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC(Br)=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results