accessibility menu, dialog, popup

UserName

CAS RN 154016-55-4

IUPAC Name: 1-(3-methyl-1,2-oxazol-5-yl)methanamine
Synonyms: 1-(3-methyl-1,2-oxazol-5-yl)methanamine
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: JKVMPILAJBLISV-UHFFFAOYSA-N
SMILES: CC1=NOC(CN)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results