accessibility menu, dialog, popup

UserName

NH2NSHNNHN

CAS RN 154-42-7

IUPAC Name: 2-amino-6,7-dihydro-3H-purine-6-thione
Synonyms: thioguanine 2-amino 6MP glaxosmithkline brand of thioguanine 2-amino 6mp tabloid thioguanine (substance) 6-mercapto-2-aminopurine 2-aminopurin-6-thiol 2-aminopurine-6(1H)-thione tabloid brand thioguanine
Molecular Weight (g/mol): 167.19
Molecular Formula: C5H5N5S
InChi Key: WYWHKKSPHMUBEB-UHFFFAOYSA-N
SMILES: NC1=NC(=S)C2=C(N1)N=CN2
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)