accessibility menu, dialog, popup

UserName

CAS RN 1539-04-4

IUPAC Name: 1,4-diphenyl benzene-1,4-dicarboxylate
Synonyms: 1,4-diphenyl benzene-1,4-dicarboxylate
Molecular Weight (g/mol): 318.33
Molecular Formula: C20H14O4
InChi Key: HPGJOUYGWKFYQW-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=C1)C1=CC=C(C=C1)C(=O)OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results