accessibility menu, dialog, popup

UserName

CAS RN 153645-26-2

CH3CH3H3COHNHOOCH3CH3H3C

CAS RN 153645-26-2

IUPAC Name: tert-butyl N-(1-hydroxy-3,3-dimethylbutan-2-yl)carbamate
Synonyms: tert-butyl N-(1-hydroxy-3,3-dimethylbutan-2-yl)carbamate
Molecular Weight (g/mol): 217.31
Molecular Formula: C11H23NO3
InChi Key: AZHJHZWFJVBGIL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CO)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)