accessibility menu, dialog, popup

UserName

CAS RN 152699-63-3

IUPAC Name: 1,3-dibenzyl 5-aminobenzene-1,3-dicarboxylate
Synonyms: 1,3-dibenzyl 5-aminobenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 361.4
Molecular Formula: C22H19NO4
InChi Key: VDJWRMQQXQBYIO-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)C(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results