accessibility menu, dialog, popup

UserName

CAS RN 152434-88-3

OHFOF

CAS RN 152434-88-3

IUPAC Name: 2,6-difluoro-3-hydroxybenzaldehyde
Synonyms: 2,6-difluoro-3-hydroxybenzaldehyde
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: NHGSYGIEJAONJB-UHFFFAOYSA-N
SMILES: OC1=C(F)C(C=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results