accessibility menu, dialog, popup

UserName

CAS RN 152044-54-7

CH3(S)(R)H3CO(S)(S)OO(S)OHCH3CH3O(R)CH3(S)OHH3CSCH3N(E)

CAS RN 152044-54-7

IUPAC Name: (1S,3S,7S,10R,11S,12S,16R)-7,11-dihydroxy-8,8,10,12,16-pentamethyl-3-[(1E)-1-(2-methyl-1,3-thiazol-4-yl)prop-1-en-2-yl]-4,17-dioxabicyclo[14.1.0]heptadecane-5,9-dione
Synonyms: epothilone B (-)-epothilone B
Molecular Weight (g/mol): 507.69
Molecular Formula: C27H41NO6S
InChi Key: QXRSDHAAWVKZLJ-PVYNADRNSA-N
SMILES: C[C@H]1CCC[C@@]2(C)O[C@H]2C[C@H](OC(=O)C[C@H](O)C(C)(C)C(=O)[C@H](C)[C@H]1O)C(\C)=C\C1=CSC(C)=N1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results