accessibility menu, dialog, popup

UserName

NH2CH2

CAS RN 1520-21-4

IUPAC Name: 4-ethenylaniline
Synonyms: benzenamine, 4-ethenyl-
Molecular Weight (g/mol): 119.17
Molecular Formula: C8H9N
InChi Key: LBSXSAXOLABXMF-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C)C=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)