accessibility menu, dialog, popup

UserName

CAS RN 151907-80-1

H3CH3CCH3OONHOHO

CAS RN 151907-80-1

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}cyclopent-2-ene-1-carboxylic acid
Synonyms: 4-[(tert-butoxycarbonyl)amino]cyclopent-2-ene-1-carboxylic acid
Molecular Weight (g/mol): 227.26
Molecular Formula: C11H17NO4
InChi Key: WOUNTSATDZJBLP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)