accessibility menu, dialog, popup

UserName

CAS RN 1515-72-6

IUPAC Name: 2-butyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: butylphthalimide
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: DLKDEVCJRCPTLN-UHFFFAOYSA-N
SMILES: CCCCN1C(=O)C2=CC=CC=C2C1=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results