CAS RN 151262-93-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
o-Flutamide, TRC
CAS: 151262-93-0 Molecular Formula: C11 H11 F3 N2 O3 Molecular Weight (g/mol): 276.21 Synonym: 2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide,Flutamide Imp. F (EP) IUPAC Name: 2-methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide SMILES: CC(C)C(=O)Nc1cc(ccc1[N+](=O)[O-])C(F)(F)F
2
2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.