CAS RN 151169-67-4
IUPAC Name:
(4-chloro-3-nitrophenyl)boronic acid
Synonyms:
4-chloro-3-nitrophenylboronic acid
Molecular Weight (g/mol):
201.37
Molecular Formula:
C6H5BClNO4
InChi Key:
LQCCRPUZTXKDGB-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-Chloro-3-nitrobenzeneboronic acid, 97%, Thermo Scientific™
CAS: 151169-67-4 Molecular Formula: C6H5BClNO4 Molecular Weight (g/mol): 201.37 MDL Number: MFCD02258950 InChI Key: LQCCRPUZTXKDGB-UHFFFAOYSA-N Synonym: 4-chloro-3-nitrophenyl boronic acid,4-chloro-3-nitrobenzeneboronic acid,3-nitro-4-chlorophenylboronic acid,5-borono-2-chloronitrobenzene,4-chloro-3-nitro-phenyl boronic acid,boronic acid,b-4-chloro-3-nitrophenyl,boronic acid, 4-chloro-3-nitrophenyl,4-chloro-3-nitrophenylboronicacid,pubchem1784,akos brn-0709 PubChem CID: 5006651 IUPAC Name: (4-chloro-3-nitrophenyl)boronic acid SMILES: OB(O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O