accessibility menu, dialog, popup

UserName

SNNNOOH

CAS RN 1506428-22-3

IUPAC Name: 4-phenyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid
Synonyms: 4-phenyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 283.31
Molecular Formula: C14H9N3O2S
InChi Key: ZCNOYYRPSQWYSP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=C(S1)C1=NC=CN=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results