CAS RN 150321-88-3
CAS RN 150321-88-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,4'-Diisothiocyano-2,2'-dihydrostilbenedisulfonic Acid Disodium Salt, TRC
CAS: 150321-88-3 Molecular Formula: C16 H10 N2 O6 S4 . 2 Na Molecular Weight (g/mol): 500.5 Synonym: 2,2'-(1,2-Ethanediyl)bis[5-isothiocyanatobenzenesulfonic Acid Disodium Salt IUPAC Name: disodium;5-isothiocyanato-2-[2-(4-isothiocyanato-2-sulfonatophenyl)ethyl]benzenesulfonate SMILES: [Na+].[Na+].[O-]S(=O)(=O)c1cc(ccc1CCc2ccc(cc2S(=O)(=O)[O-])N=C=S)N=C=S