accessibility menu, dialog, popup

UserName

CAS RN 15004-86-1

H2OH2OH2ONNNNNNClClClRh

CAS RN 15004-86-1

IUPAC Name: tris(ethane-1,2-diamine) trichlororhodium trihydrate
Synonyms: tris(ethylenediamine) trichlororhodium trihydrate
Molecular Weight (g/mol): 443.6
Molecular Formula: C6H30Cl3N6O3Rh
InChi Key: HHSLUQMEKOPKJG-UHFFFAOYSA-K
SMILES: O.O.O.NCCN.NCCN.NCCN.Cl[Rh](Cl)Cl
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)