accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 150-78-7

IUPAC Name: 1,4-dimethoxybenzene
Synonyms: 1,4-dimethoxybenzene dimethylhydroquinone P-dimethoxybenzene
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: OHBQPCCCRFSCAX-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)