accessibility menu, dialog, popup

UserName

CAS RN 150-46-9

IUPAC Name: triethyl borate
Synonyms: triethyl borate B(oet)3 triethylborat triethyl ester boric acid(H3BO3) boron triethoxide triethyl ester boric acid (H3BO3) ethyl borate ((eto)3B) boric acid(H3BO3), triethyl ester borsaeuretriethylester boric acid (H3BO3), triethyl ester
Molecular Weight (g/mol): 145.99
Molecular Formula: C6H15BO3
InChi Key: AJSTXXYNEIHPMD-UHFFFAOYSA-N
SMILES: CCOB(OCC)OCC
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)